Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543246.2 | XP_011541548.1 | 1 | 100 | ||
XM_011543248.2 | XP_011541550.1 | 1 | 185 | |||
GenBank mRNA | AJ311904.1 | - | 1 | 56 | ||
CN297963.1 | - | 1 | 203 |
Gene Symbol: ADAMTS19-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125748.1 | 1 | 221 | |||
GenBank mRNA | DA728271.1 | - | 1 | 221 |