Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTSSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138288.3 | NP_612145.2 | 1, 2 | 168 | ||
GenBank mRNA | AK001993.1 | - | 1, 2 | 149 | ||
AK315409.1 | - | 1, 2 | 146 | |||
AL080066.1 | - | 1, 2 | 149 | |||
BC016805.1 | - | 1, 2 | 150 | 4075637 | ||
BC016808.1 | - | 1, 2 | 150 | 4077762 | ||
BC021701.1 | - | 1, 2 | 149 | 4069530 | ||
BC068480.1 | - | 1, 2 | 262 | 5294549 |