Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017629.3 | NP_060099.2 | 17 | 2641 | ||
XM_005270578.3 | XP_005270635.1 | 17 | 2526 | |||
XM_005270579.3 | XP_005270636.1 | 16 | 2334 | |||
XM_011540883.2 | XP_011539185.1 | 14 | 2130 | |||
XM_011540884.2 | XP_011539186.1 | 14 | 1993 | |||
GenBank mRNA | AB046787.1 | - | 17 | 2641 | ||
AK000040.1 | - | 7 | 1130 | |||
AL831908.1 | - | 16 | 2428 | |||
BC152450.1 | - | 17 | 2641 | 8862582 |