Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017629.3 | NP_060099.2 | 2 | 279 | ||
XM_005270578.3 | XP_005270635.1 | 2 | 164 | |||
XM_017000529.1 | XP_016856018.1 | 2 | 176 | |||
GenBank mRNA | AB046787.1 | - | 2 | 279 | ||
AL831908.1 | - | 1 | 66 | |||
BC152450.1 | - | 2 | 279 | 8862582 |