Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178111.1 | NP_001171582.1 | 8 | 959 | ||
NM_001178112.1 | NP_001171583.1 | 8 | 926 | |||
NM_138967.3 | NP_620417.1 | 7 | 876 | |||
NR_033660.1 | 6 | 719 | ||||
XM_006720420.1 | XP_006720483.1 | 7 | 1035 | |||
XM_006720421.1 | XP_006720484.1 | 8 | 917 | |||
XM_006720422.3 | XP_006720485.1 | 7 | 728 | |||
XM_017021989.1 | XP_016877478.1 | 6 | 868 | |||
GenBank mRNA | AB105192.1 | - | 7 | 847 | ||
AF495715.1 | - | 7 | 828 | |||
AK056438.1 | - | 8 | 901 | |||
AK126940.1 | - | 7 | 876 | |||
AK127480.1 | - | 8 | 896 | |||
AK301505.1 | - | 6 | 681 | |||
AL833230.1 | - | 1 | 44 | |||
AL834226.1 | - | 7 | 871 | |||
BC024700.1 | - | 6 | 719 | 4941050 | ||
CR936761.1 | - | 7 | 809 |