Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300814.1 | NP_001287743.1 | 3 | 313 | ||
NM_173352.3 | NP_775487.2 | 3 | 695 | |||
XM_011538010.1 | XP_011536312.1 | 3 | 695 | |||
GenBank mRNA | AK096419.1 | - | 3 | 695 | ||
AK290901.1 | - | 3 | 695 | |||
AK298636.1 | - | 4 | 621 | |||
BX647095.1 | - | 3 | 313 |