Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182530.2 | NP_872336.2 | 17 | 2133 | ||
XM_011539052.2 | XP_011537354.1 | 16 | 2614 | |||
XM_011539055.1 | XP_011537357.1 | 15 | 1601 | |||
XM_017018967.1 | XP_016874456.1 | 16 | 2415 | |||
XM_017018968.1 | XP_016874457.1 | 17 | 2869 | |||
XM_017018969.1 | XP_016874458.1 | 16 | 2349 | |||
XM_017018970.1 | XP_016874459.1 | 15 | 2159 | |||
XM_017018971.1 | XP_016874460.1 | 17 | 2938 | |||
XM_017018972.1 | XP_016874461.1 | 17 | 2937 | |||
XM_017018973.1 | XP_016874462.1 | 13 | 1830 | |||
GenBank mRNA | AK057785.1 | - | 4 | 250 | ||
BC101820.1 | - | 2 | 85 | |||
BC143553.1 | - | 2 | 85 | 9052064 |