Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207338.3 | NP_997221.2 | 2 | 373 | ||
NR_103739.1 | 2 | 246 | ||||
XM_017021999.1 | XP_016877488.1 | 2 | 292 | |||
GenBank mRNA | AK311441.1 | - | 2 | 246 | ||
AY358729.1 | - | 2 | 373 |