Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 23 | 3025 | ||
XM_006721162.1 | XP_006721225.1 | 22 | 2955 | |||
XM_011522928.1 | XP_011521230.1 | 23 | 3203 | |||
XM_011522929.1 | XP_011521231.1 | 23 | 3068 | |||
XM_011522931.2 | XP_011521233.1 | 23 | 3090 | |||
XM_017023012.1 | XP_016878501.1 | 23 | 3052 | |||
XM_017023013.1 | XP_016878502.1 | 20 | 2848 | |||
XM_017023014.1 | XP_016878503.1 | 18 | 2980 | |||
GenBank mRNA | AJ441184.1 | - | 23 | 3024 | ||
AK056456.1 | - | 23 | 3024 | |||
AK074168.1 | - | 21 | 3556 | |||
AK092025.1 | - | 15 | 2174 | |||
AK096727.1 | - | 15 | 2231 | |||
AK123470.1 | - | 2 | 1769 | |||
AK128387.1 | - | 23 | 3043 | |||
AK315590.1 | - | 23 | 3028 | |||
AY358187.1 | - | 1 | 603 | |||
BC013735.1 | - | 21 | 2811 | |||
BC032542.1 | - | 23 | 3031 | 5534376 |