Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105513.2 | NP_001098983.2 | 21 | 2970 | ||
NM_145755.2 | NP_665698.2 | 22 | 3114 | |||
XM_005264921.4 | XP_005264978.1 | 22 | 3043 | |||
XM_005264922.4 | XP_005264979.1 | 22 | 3043 | |||
XM_005264923.3 | XP_005264980.1 | 22 | 3043 | |||
XM_005264924.4 | XP_005264981.1 | 21 | 2932 | |||
XM_005264925.4 | XP_005264982.1 | 21 | 2920 | |||
XM_005264926.4 | XP_005264983.1 | 19 | 2689 | |||
XM_005264927.4 | XP_005264984.1 | 18 | 2497 | |||
XM_006713011.3 | XP_006713074.1 | 22 | 3043 | |||
XM_011533447.2 | XP_011531749.1 | 21 | 2944 | |||
XM_017005843.1 | XP_016861332.1 | 19 | 2689 | |||
GenBank mRNA | AJ487015.1 | - | 22 | 3076 | ||
AK093313.1 | - | 12 | 1654 | |||
AK098528.1 | - | 4 | 480 | |||
AK302408.1 | - | 21 | 2953 | |||
AK302465.1 | - | 21 | 2824 | |||
BC129948.1 | - | 22 | 3025 | 40126472 | ||
BX647142.1 | - | 3 | 686 |