Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174918.2 | NP_777578.2 | 4, 5 | 398 | ||
GenBank mRNA | AF461155.1 | - | 4, 5 | 827 | ||
AF461156.1 | - | 4, 5 | 377 | |||
BC035847.1 | - | 4, 5 | 340 | |||
BC104796.1 | - | 4, 5 | 398 | 8143799 | ||
BC104798.1 | - | 4, 5 | 398 | 8143801 |