Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178134.2 | NP_835235.1 | 12 | 1386 | ||
GenBank mRNA | AK292175.1 | - | 12 | 1424 | ||
AY262056.1 | - | 12 | 1386 | |||
AY358631.1 | - | 12 | 1499 |