Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206996.2 | NP_996879.1 | 19 | 2727 | ||
XM_006710426.3 | XP_006710489.1 | 19 | 2727 | |||
XM_006710427.3 | XP_006710490.1 | 18 | 2532 | |||
XM_006710428.3 | XP_006710491.1 | 6 | 798 | |||
XM_006710429.3 | XP_006710492.1 | 3 | 410 | |||
XM_011540934.1 | XP_011539236.1 | 19 | 2727 | |||
XM_011540935.2 | XP_011539237.1 | 18 | 2566 | |||
XM_011540936.2 | XP_011539238.1 | 10 | 1385 | |||
XM_011540937.2 | XP_011539239.1 | 10 | 1551 | |||
XM_011540939.2 | XP_011539241.1 | 7 | 836 | |||
XM_011540941.2 | XP_011539243.1 | 6 | 846 | |||
XM_011540942.2 | XP_011539244.1 | 6 | 654 | |||
GenBank mRNA | AK091816.1 | - | 19 | 2713 | ||
AL833485.1 | - | 5 | 497 | |||
AY555274.1 | - | 19 | 2713 | |||
BX648110.1 | - | 19 | 2728 |