Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091902.1 | - | 3 | 558 | ||
AK124081.1 | - | 4 | 469 | |||
BC047439.1 | - | 3 | 430 | 5289095 |