Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175734.4 | NP_783861.3 | 3 | 538 | ||
XM_006721471.3 | XP_006721534.1 | 3 | 273 | |||
XM_011523722.2 | XP_011522024.1 | 3 | 1126 | |||
GenBank mRNA | BC031286.1 | - | 3 | 538 | ||
BC048010.1 | - | 3 | 538 |