Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC57 SIRNA Assays ›
Gene Symbol:
CCDC57
Gene Name:
coiled-coil domain containing 57
Gene Aliases:
-
Chromosome Location:
Chr.17: 82101458 - 82212892 on Build GRCh38
UniGene ID:
Hs.631724
Species:
Human
siRNA ID
HSS136960
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC57
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_198082.2
NP_932348.2
15
2311
XM_011523545.2
XP_011521847.2
18
2884
XM_011523546.2
XP_011521848.1
17
3216
XM_011523548.2
XP_011521850.1
18
2912
XM_011523549.2
XP_011521851.1
17
2726
XM_011523551.2
XP_011521853.1
18
2925
XM_011523552.2
XP_011521854.1
16
2284
XM_011523555.2
XP_011521857.1
15
3284
XM_011523557.2
XP_011521859.1
11
1648
XM_017024462.1
XP_016879951.1
18
2886
XM_017024463.1
XP_016879952.1
18
2875
XM_017024464.1
XP_016879953.1
18
2877
XM_017024465.1
XP_016879954.1
18
2876
XM_017024466.1
XP_016879955.1
18
2865
XM_017024467.1
XP_016879956.1
18
3041
XM_017024468.1
XP_016879957.1
18
2872
XM_017024469.1
XP_016879958.1
18
2704
XM_017024470.1
XP_016879959.1
17
2505
XM_017024471.1
XP_016879960.1
17
2681
XM_017024472.1
XP_016879961.1
18
2902
XM_017024473.1
XP_016879962.1
17
2657
XM_017024474.1
XP_016879963.1
17
2753
XM_017024475.1
XP_016879964.1
18
2611
XM_017024476.1
XP_016879965.1
16
2787
XM_017024477.1
XP_016879966.1
17
2955
XM_017024478.1
XP_016879967.1
15
2418
XM_017024479.1
XP_016879968.1
17
3091
XM_017024483.1
XP_016879972.1
15
2520
GenBank mRNA
AK074334.1
-
3
209
BC110997.1
-
16
2314
5725904
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline