Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159770.1 | NP_001153242.1 | 7 | 749 | ||
NM_139177.3 | NP_631916.2 | 7 | 728 | |||
XM_005257134.3 | XP_005257191.1 | 7 | 815 | |||
XM_006721754.3 | XP_006721817.1 | 7 | 836 | |||
XM_006721755.2 | XP_006721818.1 | 8 | 1040 | |||
XM_006721756.2 | XP_006721819.1 | 7 | 749 | |||
XM_006721757.2 | XP_006721820.1 | 6 | 578 | |||
XM_006721759.1 | XP_006721822.1 | 7 | 749 | |||
XM_011524493.1 | XP_011522795.1 | 8 | 1131 | |||
XM_011524494.2 | XP_011522796.1 | 7 | 782 | |||
XM_011524495.2 | XP_011522797.1 | 7 | 749 | |||
XM_017024330.1 | XP_016879819.1 | 8 | 1019 | |||
XM_017024331.1 | XP_016879820.1 | 8 | 1110 | |||
XM_017024332.1 | XP_016879821.1 | 7 | 725 | |||
XM_017024333.1 | XP_016879822.1 | 6 | 689 | |||
XM_017024334.1 | XP_016879823.1 | 6 | 665 | |||
XM_017024335.1 | XP_016879824.1 | 6 | 557 | |||
XM_017024336.1 | XP_016879825.1 | 8 | 929 | |||
XM_017024337.1 | XP_016879826.1 | 8 | 842 | |||
XM_017024342.1 | XP_016879831.1 | 7 | 728 | |||
GenBank mRNA | AF331643.1 | - | 7 | 690 | ||
AK095227.1 | - | 7 | 723 | |||
AK313376.1 | - | 7 | 701 | |||
BC035631.1 | - | 7 | 707 | 5219031 |