Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256124.1 | NP_001243053.1 | 5 | 1023 | ||
NM_173547.3 | NP_775818.2 | 6 | 1089 | |||
XM_006721760.3 | XP_006721823.1 | 6 | 1126 | |||
XM_006721761.3 | XP_006721824.1 | 5 | 1060 | |||
XM_011524499.2 | XP_011522801.1 | 6 | 1129 | |||
XM_011524500.2 | XP_011522802.1 | 6 | 1130 | |||
XM_011524501.2 | XP_011522803.1 | 5 | 1064 | |||
XM_011524502.2 | XP_011522804.1 | 5 | 1064 | |||
XM_011524503.2 | XP_011522805.1 | 4 | 892 | |||
XM_017024343.1 | XP_016879832.1 | 5 | 958 | |||
XM_017024344.1 | XP_016879833.1 | 4 | 850 | |||
GenBank mRNA | BC006138.1 | - | 3 | 198 | ||
BC021259.2 | - | 6 | 1057 | 4128330 | ||
BC073831.1 | - | 3 | 287 | |||
BC098412.1 | - | 13 | ||||
BG753016.1 | - | 5 | 579 |