Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322176.1 | NP_001309105.1 | 1 | 823 | ||
NM_001322177.1 | NP_001309106.1 | 1 | 823 | |||
NM_177966.6 | NP_808881.3 | 1 | 823 | |||
GenBank mRNA | AB115695.1 | - | 1 | 761 | ||
AK074423.1 | - | 1 | 772 | |||
AK300374.1 | - | 1 | 796 | |||
AL831824.1 | - | 1 | 327 | |||
BX647169.1 | - | 1 | 793 |