Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152678.2 | NP_689891.1 | 17, 18 | 1596 | ||
XM_006713019.3 | XP_006713082.1 | 17, 18 | 1622 | |||
XM_006713020.2 | XP_006713083.1 | 18, 19 | 1828 | |||
XM_017005864.1 | XP_016861353.1 | 18, 19 | 1831 | |||
GenBank mRNA | AK074156.1 | - | 17, 18 | 1583 | ||
AK092288.1 | - | 12, 13 | 998 | |||
BC040291.1 | - | 17, 18 | 1584 | 4822560 | ||
BC054343.1 | - | 8, 9 | 679 |