Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152680.2 | NP_689893.1 | 2 | 473 | ||
XM_011531716.2 | XP_011530018.1 | 2 | 331 | |||
GenBank mRNA | AK056590.1 | - | 2 | 347 | ||
AK296694.1 | - | 2 | 298 | |||
AK309319.1 | - | 2 | 320 | |||
BC019600.1 | - | 1 | 76 | |||
BC060775.1 | - | 2 | 245 | 30336271 |