Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317896.1 | NP_001304825.1 | 3 | 453 | ||
NM_001317897.1 | NP_001304826.1 | 2 | 162 | |||
NM_174921.2 | NP_777581.1 | 2 | 308 | |||
XM_011513640.2 | XP_011511942.1 | 2 | 441 | |||
XM_017007862.1 | XP_016863351.1 | 3 | 357 | |||
XM_017007863.1 | XP_016863352.1 | 2 | 198 | |||
GenBank mRNA | AK023351.1 | - | 2 | 171 | ||
AK307146.1 | - | 2 | 201 | |||
BC008502.1 | - | 2 | 171 | 4295121 | ||
BQ049060.1 | - | 2 | 153 | |||
HY004916.1 | - | 3 | 354 | |||
HY155982.1 | - | 2 | 309 |