Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153692.3 | NP_710159.1 | 3 | 857 | ||
XM_011544431.2 | XP_011542733.1 | 3 | 857 | |||
XM_011544432.1 | XP_011542734.1 | 3 | 857 | |||
GenBank mRNA | AK075205.1 | - | 3 | 828 | ||
AK300197.1 | - | 3 | 736 | |||
BC057765.1 | - | 3 | 789 | 30348811 |