Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313994.1 | NP_001300923.1 | 16, 17 | 2010 | ||
NM_145004.6 | NP_659441.3 | 19, 20 | 2408 | |||
XM_006716298.2 | XP_006716361.1 | 19, 20 | 2202 | |||
XM_006716299.1 | XP_006716362.1 | 17, 18 | 1884 | |||
XM_011544433.1 | XP_011542735.1 | 19, 20 | 2202 | |||
XM_011544434.1 | XP_011542736.1 | 17, 18 | 1983 | |||
XM_017013183.1 | XP_016868672.1 | 18, 19 | 2133 | |||
XM_017013185.1 | XP_016868674.1 | 18, 19 | 2208 | |||
XM_017013186.1 | XP_016868675.1 | 15, 16 | 1812 | |||
XM_017013187.1 | XP_016868676.1 | 11, 12 | 1348 | |||
XM_017013188.1 | XP_016868677.1 | 10, 11 | 1317 | |||
GenBank mRNA | AK122608.1 | - | 17, 18 | 1763 | ||
AK131038.1 | - | 11, 12 | 1377 | |||
AK300148.1 | - | 16, 17 | 1968 | |||
AK313864.1 | - | 19, 20 | 2229 | |||
AY358739.1 | - | 19, 20 | 2288 | |||
BC026085.2 | - | 19, 20 | 2192 | 4838404 | ||
BC026169.2 | - | 19, 20 | 2219 | 4829390 | ||
BC028702.2 | - | 19, 20 | 2202 | 4829385 | ||
BC030014.2 | - | 19, 20 | 2195 | 4830659 | ||
BC030698.2 | - | 19, 20 | 2210 | 4822988 | ||
BC034975.2 | - | 19, 20 | 2203 | 4829806 |