Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173550.2 | NP_775821.2 | 6 | 892 | ||
XM_005251397.2 | XP_005251454.1 | 6 | 775 | |||
XM_005251398.3 | XP_005251455.1 | 6 | 1014 | |||
XM_005251399.4 | XP_005251456.1 | 7 | 1074 | |||
XM_011517788.2 | XP_011516090.1 | 6 | 1004 | |||
XM_011517796.2 | XP_011516098.1 | 4 | 421 | |||
XM_017014431.1 | XP_016869920.1 | 6 | 1014 | |||
XM_017014432.1 | XP_016869921.1 | 6 | 1014 | |||
XM_017014433.1 | XP_016869922.1 | 6 | 1014 | |||
XM_017014434.1 | XP_016869923.1 | 6 | 1014 | |||
XM_017014435.1 | XP_016869924.1 | 6 | 1014 | |||
XM_017014436.1 | XP_016869925.1 | 6 | 1014 | |||
XM_017014437.1 | XP_016869926.1 | 6 | 1014 | |||
XM_017014438.1 | XP_016869927.1 | 6 | 1013 | |||
XM_017014439.1 | XP_016869928.1 | 7 | 1176 | |||
XM_017014440.1 | XP_016869929.1 | 4 | 435 | |||
XM_017014441.1 | XP_016869930.1 | 6 | 1013 | |||
XM_017014442.1 | XP_016869931.1 | 6 | 1013 | |||
XM_017014443.1 | XP_016869932.1 | 4 | 416 | |||
GenBank mRNA | AK308284.1 | - | 6 | 775 | ||
AY422473.1 | - | 6 | 892 | |||
BC144214.1 | - | 5 | 669 | 9052734 | ||
BC150600.1 | - | 5 | 669 | 9056970 | ||
BX537933.1 | - | 6 | 753 | |||
CR936775.1 | - | 7 | 1146 |