Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145007.3 | NP_659444.2 | 4 | 877 | ||
GenBank mRNA | AK310394.1 | - | 4 | 554 | ||
AY095145.1 | - | 2 | 265 | |||
AY154466.1 | - | 1 | 166 | |||
AY168967.1 | - | 2 | 254 | |||
AY226382.1 | - | 1 | 166 | |||
BC034730.1 | - | 2 | 254 | 4341197 |