Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152696.3 | NP_689909.2 | 6, 7 | 1750 | ||
NM_181358.2 | NP_852003.1 | 4, 5 | 470 | |||
NM_198268.2 | NP_938009.1 | 6, 7 | 1750 | |||
NM_198269.2 | NP_938010.1 | 5, 6 | 661 | |||
XM_005270609.3 | XP_005270666.1 | 6, 7 | 1927 | |||
XM_005270610.4 | XP_005270667.1 | 6, 7 | 1927 | |||
XM_005270611.4 | XP_005270668.1 | 6, 7 | 1927 | |||
XM_005270612.4 | XP_005270669.1 | 6, 7 | 1889 | |||
XM_005270613.4 | XP_005270670.1 | 8, 9 | 1840 | |||
XM_006710443.3 | XP_006710506.1 | 6, 7 | 1927 | |||
XM_011540977.2 | XP_011539279.1 | 5, 6 | 1807 | |||
XM_017000605.1 | XP_016856094.1 | 6, 7 | 1889 | |||
GenBank mRNA | AB089957.1 | - | 6, 7 | 1714 | ||
AK122662.1 | - | 6, 7 | 1748 | |||
AK302726.1 | - | 6, 7 | 1934 | |||
AL833829.1 | - | 5, 6 | 639 | |||
BC028408.1 | - | 5, 6 | 666 | 4821915 | ||
BC033012.2 | - | 6, 7 | 1750 | 4830408 | ||
BC036057.1 | - | 4, 5 | 470 | 5272816 |