Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LVRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173800.4 | NP_776161.3 | 14 | 2281 | ||
GenBank mRNA | AK075131.1 | - | 9 | 816 | ||
AK291655.1 | - | 14 | 2295 | |||
AY560010.1 | - | 14 | 2169 | |||
BC036440.1 | - | 9 | 1160 | |||
BC060869.1 | - | 15 | 2208 | 30346049 | ||
BC070028.1 | - | 9 | 1278 | |||
BC094716.1 | - | 9 | 1278 | |||
BC109022.1 | - | 14 | 2145 | 40008501 | ||
BC109023.1 | - | 14 | 2145 | 40008502 |