Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRSX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145177.2 | NP_660160.2 | 2, 3 | 258 | ||
GenBank mRNA | AJ293620.1 | - | 2, 3 | 218 | ||
AY358849.1 | - | 2, 3 | 220 | |||
BC019696.2 | - | 2, 3 | 211 | |||
BC032340.1 | - | 2, 3 | 222 | 5192975 |