Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130047.1 | NP_001123519.1 | 5 | 702 | ||
NM_152715.3 | NP_689928.3 | 5 | 829 | |||
XM_017017326.1 | XP_016872815.1 | 6 | 928 | |||
GenBank mRNA | AK295673.1 | - | 5 | 820 | ||
AK313324.1 | - | 5 | 724 | |||
AY398644.1 | - | 5 | 812 | |||
BC020501.1 | - | 5 | 738 | |||
BC120988.1 | - | 4 | 652 | 40116354 |