Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098835.1 | NP_001092305.1 | 6 | 781 | ||
NM_001278242.1 | NP_001265171.1 | 5 | 658 | |||
NM_152717.2 | NP_689930.1 | 5 | 494 | |||
NR_103481.1 | 7 | 1068 | ||||
XM_006718459.3 | XP_006718522.1 | 5 | 501 | |||
XM_011544812.2 | XP_011543114.1 | 6 | 745 | |||
XM_011544814.1 | XP_011543116.1 | 6 | 610 | |||
GenBank mRNA | AK091846.1 | - | 7 | 931 | ||
AK300233.1 | - | 5 | 572 | |||
AY584608.1 | - | 6 | 640 | |||
AY584609.1 | - | 5 | 517 | |||
BC031610.1 | - | 5 | 358 | 5176452 |