Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182833.1 | NP_878253.1 | 11 | 1200 | ||
XM_011544826.1 | XP_011543128.1 | 11 | 2931 | |||
XM_011544833.1 | XP_011543135.1 | 11 | 2931 | |||
XM_011544834.1 | XP_011543136.1 | 12 | 1426 | |||
XM_017017348.1 | XP_016872837.1 | 12 | 1370 | |||
XM_017017349.1 | XP_016872838.1 | 11 | 1287 | |||
XM_017017350.1 | XP_016872839.1 | 11 | 2802 | |||
XM_017017351.1 | XP_016872840.1 | 11 | 1158 | |||
GenBank mRNA | AY313782.1 | - | 11 | 1200 | ||
AY326450.1 | - | 11 | 1200 |