Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286159.1 | NP_001273088.1 | 4, 5 | 863 | ||
NM_173556.4 | NP_775827.2 | 4, 5 | 863 | |||
XM_011544839.2 | XP_011543141.1 | 4, 5 | 449 | |||
XM_011544840.2 | XP_011543142.1 | 4, 5 | 521 | |||
XM_011544841.1 | XP_011543143.1 | 4, 5 | 411 | |||
XM_011544842.1 | XP_011543144.1 | 4, 5 | 863 | |||
XM_011544844.2 | XP_011543146.1 | 2, 3 | 575 | |||
XM_017017354.1 | XP_016872843.1 | 4, 5 | 863 | |||
GenBank mRNA | AK093405.1 | - | 2, 3 | 243 | ||
AK124113.1 | - | 4, 5 | 864 | |||
AK314036.1 | - | 4, 5 | 567 | |||
BC040280.1 | - | 4, 5 | 541 | 5166625 |