Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205138.3 | NP_001192067.1 | 5 | 1119 | ||
NM_001271471.2 | NP_001258400.1 | 5 | 1538 | |||
NM_001318803.1 | NP_001305732.1 | 6 | 1213 | |||
NM_145309.5 | NP_660352.1 | 6 | 1256 | |||
NR_026886.3 | 7 | 1436 | ||||
NR_134858.1 | 4 | 1035 | ||||
XM_006718473.3 | XP_006718536.1 | 6 | 1482 | |||
XM_006718474.3 | XP_006718537.1 | 5 | 1521 | |||
XM_011544847.2 | XP_011543149.1 | 6 | 1710 | |||
XM_011544848.2 | XP_011543150.1 | 7 | 1768 | |||
XM_017017359.1 | XP_016872848.1 | 7 | 1873 | |||
XM_017017360.1 | XP_016872849.1 | 7 | 1725 | |||
GenBank mRNA | AK298997.1 | - | 5 | 638 | ||
AK311690.1 | - | 7 | 906 | |||
AK313150.1 | - | 6 | 940 | |||
AK315892.1 | - | 3 | 334 | |||
BC012855.1 | - | 6 | 728 | 3863323 | ||
BI461207.1 | - | 6 | 699 | |||
EU627066.1 | - | 6 | 728 | |||
EU627068.1 | - | 5 | 1009 |