Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286341.1 | NP_001273270.1 | 1 | 147 | ||
NM_152719.2 | NP_689932.1 | 1 | 147 | |||
XM_011534971.2 | XP_011533273.1 | 1 | 1421 | |||
GenBank mRNA | AK093129.1 | - | 1 | 140 | ||
HY007057.1 | - | 1 | 139 | |||
HY017873.1 | - | 1 | 149 | |||
HY026582.1 | - | 1 | 31 |