Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039535.2 | NP_001034624.1 | 3, 4 | 452 | ||
NM_145060.3 | NP_659497.1 | 3, 4 | 407 | |||
GenBank mRNA | AK303464.1 | - | 3, 4 | 386 | ||
AK313968.1 | - | 3, 4 | 434 | |||
BC015706.1 | - | 3, 4 | 396 | 3909951 | ||
BG493245.1 | - | 3, 4 | 427 | |||
BM470268.1 | - | 147 | ||||
BQ069178.1 | - | 508 | ||||
BX956145.1 | - | 3, 4 | 355 | |||
CN403719.1 | - | 3, 4 | 305 |