Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039535.2 | NP_001034624.1 | 6 | 806 | ||
NM_145060.3 | NP_659497.1 | 6 | 761 | |||
GenBank mRNA | AK303464.1 | - | 5 | 602 | ||
AK313968.1 | - | 6 | 788 | |||
BC015706.1 | - | 6 | 750 | 3909951 | ||
BM470268.1 | - | 501 | ||||
CN403719.1 | - | 6 | 659 |