Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323977.1 | NP_001310906.1 | 2 | 156 | ||
NM_001323978.1 | NP_001310907.1 | 3 | 584 | |||
NM_001323979.1 | NP_001310908.1 | 2 | 271 | |||
NM_001323980.1 | NP_001310909.1 | 2 | 501 | |||
NM_001323981.1 | NP_001310910.1 | 3 | 714 | |||
NM_194298.2 | NP_919274.1 | 2 | 631 | |||
XM_017015883.1 | XP_016871372.1 | 2 | 631 | |||
XM_017015884.1 | XP_016871373.1 | 3 | 964 | |||
GenBank mRNA | AK125791.1 | - | 2 | 291 | ||
BC126238.1 | - | 1 | 220 | 8991954 | ||
BC130567.1 | - | 1 | 220 | 40146598 | ||
DA759951.1 | - | 2 | 156 |