Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318140.1 | NP_001305069.1 | 5 | 489 | ||
NM_001318141.1 | NP_001305070.1 | 5 | 467 | |||
NM_001318142.1 | NP_001305071.1 | 4 | 345 | |||
NM_001318143.1 | NP_001305072.1 | 3 | 249 | |||
NM_145312.3 | NP_660355.2 | 5 | 472 | |||
XM_011539498.2 | XP_011537800.1 | 3 | 1808 | |||
GenBank mRNA | AK074679.1 | - | 5 | 472 | ||
AK299707.1 | - | 3 | 249 | |||
AK313328.1 | - | 4 | 301 | |||
BC014161.2 | - | 5 | 470 | 4543606 | ||
BX648149.1 | - | 5 | 418 | |||
DB448752.1 | - | 4 | 289 | |||
DN994938.1 | - | 5 | 461 |