Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282862.1 | NP_001269791.1 | 2 | 286 | ||
NM_145313.3 | NP_660356.2 | 2 | 258 | |||
GenBank mRNA | AK095136.1 | - | 2 | 286 | ||
BC022548.1 | - | 2 | 258 | 4814750 | ||
HY118234.1 | - | 2 | 259 |