Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166271.2 | NP_001159743.1 | 4, 5 | 2481 | ||
NM_001286792.1 | NP_001273721.1 | 6, 7 | 2805 | |||
NM_153023.3 | NP_694568.1 | 3, 4 | 717 | |||
GenBank mRNA | AK055770.1 | - | 3, 4 | 685 | ||
AK299981.1 | - | 1, 2 | 158 | |||
AK301149.1 | - | 1, 2 | 158 | |||
BC109290.1 | - | 2, 3 | 427 | 40034357 | ||
BC109291.1 | - | 2, 3 | 427 | 40034359 | ||
BX648244.1 | - | 6, 7 | 2627 |