Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143685.1 | NP_001137157.1 | 10 | 1401 | ||
NM_001190158.1 | NP_001177087.1 | 11 | 1485 | |||
NM_145024.2 | NP_659461.1 | 10 | 1401 | |||
GenBank mRNA | AK056109.1 | - | 10 | 1401 | ||
AK090997.1 | - | 10 | 1327 | |||
AK294334.1 | - | 11 | 1485 | |||
AY907669.1 | - | 10 | 1328 | |||
BC039073.1 | - | 10 | 1414 | |||
BC069501.1 | - | 10 | 1372 | 7262427 | ||
BC069548.1 | - | 10 | 1371 | 7262439 | ||
BC117126.1 | - | 10 | 1374 | 40125677 | ||
BC143692.1 | - | 10 | 1374 | 9052206 | ||
EU794678.1 | - | 10 | 1401 |