Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 11 | 1280 | ||
GenBank mRNA | AK090918.1 | - | 8 | 2010 | ||
AK290408.1 | - | 11 | 1230 | |||
AK308337.1 | - | 11 | 1264 | |||
AY203935.1 | - | 8 | 891 | |||
BC015138.1 | - | 10 | 1060 | |||
BC108699.1 | - | 11 | 1237 | 6061738 | ||
BC136832.1 | - | 11 | 1239 | 9020823 | ||
BC136841.1 | - | 11 | 1239 | 9020832 | ||
CA390074.1 | - | 1 | 69 |