Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 7 | 720 | ||
XM_006715378.3 | XP_006715441.1 | 7 | 723 | |||
GenBank mRNA | AK090918.1 | - | 4 | 1450 | ||
AK290408.1 | - | 7 | 670 | |||
AK308337.1 | - | 7 | 703 | |||
AY203935.1 | - | 4 | 330 | |||
BC015138.1 | - | 6 | 500 | |||
BC108699.1 | - | 7 | 677 | 6061738 | ||
BC136832.1 | - | 7 | 679 | 9020823 | ||
BC136841.1 | - | 7 | 679 | 9020832 | ||
BM808222.1 | - | 711 |