Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM26D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256887.2 | NP_001243816.1 | 4 | 677 | ||
NM_001256888.2 | NP_001243817.1 | 4 | 583 | |||
NM_001256889.2 | NP_001243818.1 | 3 | 461 | |||
NM_153036.4 | NP_694581.1 | 3 | 516 | |||
XM_005266860.3 | XP_005266917.1 | 2 | 1451 | |||
XM_011535560.2 | XP_011533862.1 | 5 | 771 | |||
XM_011535561.2 | XP_011533863.1 | 4 | 620 | |||
XM_011535562.2 | XP_011533864.1 | 6 | 952 | |||
XM_011535563.2 | XP_011533865.1 | 4 | 565 | |||
XM_011535564.1 | XP_011533866.1 | 4 | 526 | |||
XM_017010390.1 | XP_016865879.1 | 5 | 699 | |||
XM_017010391.1 | XP_016865880.1 | 5 | 654 | |||
XM_017010392.1 | XP_016865881.1 | 5 | 716 | |||
GenBank mRNA | AK056801.1 | - | 3 | 449 | ||
AK300309.1 | - | 4 | 585 | |||
AY358252.1 | - | 3 | 498 | |||
BC057769.1 | - | 4 | 646 | 30332648 | ||
DA855717.1 | - | 3 | 428 |