Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098518.1 | NP_001091988.1 | 17 | 3368 | ||
NM_015234.4 | NP_056049.4 | 17 | 3308 | |||
XM_005248892.2 | XP_005248949.1 | 18 | 3488 | |||
XM_005248893.3 | XP_005248950.1 | 18 | 3444 | |||
XM_005248894.3 | XP_005248951.1 | 18 | 3488 | |||
XM_005248895.3 | XP_005248952.1 | 18 | 3488 | |||
XM_011514353.2 | XP_011512655.1 | 17 | 3368 | |||
XM_011514354.2 | XP_011512656.1 | 19 | 3511 | |||
XM_017010418.1 | XP_016865907.1 | 18 | 3444 | |||
XM_017010419.1 | XP_016865908.1 | 17 | 3324 | |||
XM_017010420.1 | XP_016865909.1 | 17 | 3368 | |||
XM_017010421.1 | XP_016865910.1 | 17 | 3324 | |||
XM_017010422.1 | XP_016865911.1 | 17 | 3284 | |||
XM_017010423.1 | XP_016865912.1 | 18 | 3391 | |||
XM_017010424.1 | XP_016865913.1 | 18 | 3391 | |||
GenBank mRNA | AB018301.2 | - | 17 | 3306 | ||
AB031421.1 | - | 16 | 3123 | |||
AK075121.1 | - | 1 | 256 | |||
AK289503.1 | - | 17 | 3306 | |||
AK296948.1 | - | 7 | 1691 | |||
AK300380.1 | - | 6 | 1879 | |||
AL832125.1 | - | 15 | 2849 | |||
AY140958.1 | - | 16 | 3076 | |||
AY255552.1 | - | 1 | 235 | |||
AY498875.1 | - | 17 | 3167 | |||
BC050087.1 | - | 4 | 1192 | |||
BC066121.1 | - | 17 | 3146 | 5298732 |