Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162900.1 | NP_001156372.1 | 2 | 1128 | ||
NM_001286401.1 | NP_001273330.1 | 2 | 1123 | |||
NM_145316.3 | NP_660359.2 | 2 | 605 | |||
XM_011514362.1 | XP_011512664.1 | 2 | 591 | |||
XM_011514363.2 | XP_011512665.1 | 2 | 591 | |||
XM_011514364.2 | XP_011512666.1 | 2 | 1375 | |||
XM_011514365.2 | XP_011512667.1 | 2 | 1160 | |||
XM_011514366.2 | XP_011512668.1 | 2 | 591 | |||
XM_011514367.2 | XP_011512669.1 | 2 | 591 | |||
GenBank mRNA | AK098666.1 | - | 2 | 564 | ||
BC009625.1 | - | 2 | 1101 | |||
BC026012.1 | - | 2 | 562 | 5269554 |