Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173558.3 | NP_775829.2 | 14 | 1668 | ||
XM_017010429.1 | XP_016865918.1 | 15 | 1681 | |||
GenBank mRNA | AK092732.1 | - | 13 | 1877 | ||
AK097230.1 | - | 2 | 868 | |||
AK097981.1 | - | 10 | 1552 | |||
AK131079.1 | - | 8 | 4918 | |||
AK290419.1 | - | 15 | 1661 | |||
AK301156.1 | - | 12 | 1401 | |||
BC023645.2 | - | 14 | 1668 | 5087501 | ||
BC053655.1 | - | 15 | 1613 | 5755893 | ||
BX648164.1 | - | 14 | 1572 |