Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199159.1 | NP_001186088.1 | 6 | 840 | ||
NM_153370.2 | NP_699201.2 | 5 | 945 | |||
XM_005248917.2 | XP_005248974.1 | 5 | 774 | |||
XM_011514375.2 | XP_011512677.1 | 5 | 774 | |||
XM_017010430.1 | XP_016865919.1 | 5 | 774 | |||
GenBank mRNA | AK075470.1 | - | 5 | 640 | ||
AK124589.1 | - | 8 | 2464 | |||
AK303928.1 | - | 4 | 594 | |||
AK312785.1 | - | 5 | 638 | |||
AY358422.1 | - | 5 | 631 | |||
BC022399.1 | - | 5 | 639 | 4691115 | ||
BC035634.1 | - | 5 | 945 | 5217121 |