Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111038.1 | NP_001104508.1 | 19 | 2147 | ||
NM_001287150.1 | NP_001274079.1 | 19 | 2198 | |||
NM_145032.3 | NP_659469.3 | 20 | 2459 | |||
NR_105043.1 | 21 | 2296 | ||||
XM_005250205.3 | XP_005250262.1 | 21 | 2365 | |||
XM_005250207.3 | XP_005250264.1 | 20 | 2266 | |||
XM_005250208.3 | XP_005250265.1 | 20 | 2230 | |||
XM_005250209.2 | XP_005250266.1 | 20 | 2263 | |||
XM_006715898.2 | XP_006715961.1 | 15 | 1710 | |||
XM_011515928.2 | XP_011514230.1 | 20 | 2281 | |||
XM_011515930.2 | XP_011514232.1 | 20 | 2329 | |||
XM_017011850.1 | XP_016867339.1 | 21 | 2309 | |||
XM_017011852.1 | XP_016867341.1 | 21 | 2515 | |||
XM_017011853.1 | XP_016867342.1 | 20 | 2315 | |||
GenBank mRNA | AK095387.1 | - | 15 | 1682 | ||
AK097537.1 | - | 20 | 2459 | |||
AL713709.1 | - | 11 | 1181 | |||
AY359238.1 | - | 13 | 1320 | |||
AY359239.1 | - | 19 | 2031 | |||
BC020572.2 | - | 14 | 1536 | |||
BC020575.2 | - | 6 | 569 | |||
BC026121.1 | - | 21 | 2299 | 4826546 | ||
BC031285.1 | - | 19 | 2190 | 5298780 | ||
BP378950.1 | - | 1 | 21 |